Cable lockouts, double looped
Leverantör: Brady
BRDY131063EA 165 SEK
BRDY131063 BRDY131064 BRDY131065 BRDY131066
Cable lockouts, double looped
Säkerhetslås
Double looped cable lockout devices made from plastic coated steel are ideal for locking out large valves in many different industries. For any situation that requires a flexible lockout device and a visual deterrent.
  • Non conductive
  • Heavy duty cables
  • Come in four lengths to ensure a secure fit
Many customers use large, heavy chains to lock out valves. These double looped cables are much more lightweight and are better visual deterrent that the isolation point is locked.
Cable diameter: 4,76 mm