Anti-CCNB1 Rabbit Polyclonal Antibody

Leverantör: ProSci Inc.
PRSI27-300EA 6850 SEK
PRSI27-300
Anti-CCNB1 Rabbit Polyclonal Antibody
Antikroppar
CCNB1 is a regulatory protein involved in mitosis. CCNB1 complexes with p34 (cdc2) to form the maturation-promoting factor (MPF).The protein encoded by this gene is a regulatory protein involved in mitosis. The gene product complexes with p34 (cdc2) to form the maturation-promoting factor (MPF). Two alternative transcripts have been found, a constitutively expressed transcript and a cell cycle-regulated transcript, that is expressed predominantly during G2/M phase. The different transcripts result from the use of alternate transcription initiation sites.

CCNB1 antibody can be used for detection of CCNB1 by ELISA at 1:312500. CCNB1 antibody can be used for detection of CCNB1 by western blot at 0.125 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CCNB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR