Håll mig inloggad?
Avslutas om 
Din inloggning har gått ut
Din session har gått ut. För din säkerhet har vi loggat ut dig.
Vill du logga in igen?

Anti-HSPB1 Rabbit Polyclonal Antibody

Leverantör: ProSci Inc.
HSPB1, like the other heat shock proteins, is part of a complex system of molecular chaperones in epidermal keratinocytes.

HSPB1 antibody can be used for detection of HSPB1 by ELISA at 1:62500. HSPB1 antibody can be used for detection of HSPB1 by western blot at 1.25 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: HSPB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human




 

ORDER

Expandera alla / Komprimera alla
Storlek Förp. VWR art.nr Antal
100 µg 100 µG PRSI27-301
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFIKATIONER

Antigen symbol HSPB1
Antigen name Heat Shock 27kDa Protein 1
Reaktivitet Human
Conjugation Unconjugated
Clonality Polyclonal
Värd Rabbit
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Antigen synonymer HMN2B, SRP27, CMT2F, HS.76067, HSP27, Hsp25, HSP28, HEL-S-102
Immunogen A synthetic peptide corresponding a region of human HSPB1.
Form Lyophilized
Enter Details

Select the article for which you are looking for Certificate

Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping

Försäljningsbestämmelser

All försäljning och leverans sker enligt de allmänna leveransbestämmelserna IML 2009 (antagna 2008 av Swedish Labtech) med tillägg av vid var tid tillämpliga försäljningsbestämmelser för VWR, som ska gälla med företräde framför IML 2009 i händelse av motsättning.