Håll mig inloggad?
Avslutas om 
Din inloggning har gått ut
Din session har gått ut. För din säkerhet har vi loggat ut dig.
Vill du logga in igen?

Anti-POU6F1 Rabbit Polyclonal Antibody

Leverantör: ProSci Inc.
The POU6F1 gene encodes a protein that is part of a family of transcription factors which exhibit distinct temporal and spatial patterns of expression.

POU6F1 antibody can be used for detection of POU6F1 by ELISA at 1:312500. POU6F1 antibody can be used for detection of POU6F1 by western blot at 0.12 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: POU6F1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse, Rat




 

ORDER

Expandera alla / Komprimera alla
Storlek Förp. VWR art.nr Antal
50 µg 50 µG PRSI27-341
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFIKATIONER

Antigen symbol POU6F1
Antigen name POU Domain Class 6, Transcription Factor 1
Reaktivitet Mouse, Rat, Dog, Human
Conjugation Unconjugated
Clonality Polyclonal
Värd Rabbit
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Antigen synonymer TCFB1, MPOU, BRN5
Immunogen A synthetic peptide corresponding a region of human POU6F1.
Form Lyophilized
Enter Details

Select the article for which you are looking for Certificate

Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal