Anti-PQBP1 Rabbit Polyclonal Antibody

Leverantör: ProSci Inc.
PRSI27-351EA 6250 SEK
PRSI27-351
Anti-PQBP1 Rabbit Polyclonal Antibody
Antikroppar
PQBP1 is a nuclear polyglutamine-binding protein that contains a WW domain. Mutations in this gene are associated with X-linked mental retardation.

PQBP1 antibody can be used for detection of PQBP1 by ELISA at 1:312500. PQBP1 antibody can be used for detection of PQBP1 by western blot at 2.5 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PQBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR