Håll mig inloggad?
Avslutas om 
Din inloggning har gått ut
Din session har gått ut. För din säkerhet har vi loggat ut dig.
Vill du logga in igen?

Anti-YAF2 Rabbit Polyclonal Antibody

Leverantör: ProSci Inc.
YAF2 interacts with YY1, a zinc finger protein involved in negative regulation of muscle-restricted genes. YAF2 contains a single N-terminal C2-X10-C2 zinc finger, and in contrast to YY1, is up-regulated during myogenic differentiation. It also facilitates proteolytic cleavage of YY1 by the calcium- activated protease, m-calpain, suggesting a mechanism by which this protein antagonizes the negative effect of YY1.The protein encoded by this gene interacts with YY1, a zinc finger protein involved in negative regulation of muscle-restricted genes. This gene product itself contains a single N-terminal C2-X10-C2 zinc finger, and in contrast to YY1, is up-regulated during myogenic differentiation. It also facilitates proteolytic cleavage of YY1 by the calcium- activated protease, m-calpain, suggesting a mechanism by which this protein antagonizes the negative effect of YY1. Multiple transcript variants encoding different isoforms have been found for this gene, but the full-length nature of only two have been confirmed to date.

YAF2 antibody can be used for detection of YAF2 by ELISA at 1:62500. YAF2 antibody can be used for detection of YAF2 by western blot at 1.25 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: YAF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human




 

ORDER

Expandera alla / Komprimera alla
Storlek Förp. VWR art.nr Antal
100 µg 100 µG PRSI27-352
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFIKATIONER

Antigen symbol YAF2
Reaktivitet Human, Dog
Conjugation Unconjugated
Clonality Polyclonal
Värd Rabbit
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Immunogen A synthetic peptide corresponding a region of human YAF2.
Form Lyophilized
Enter Details

Select the article for which you are looking for Certificate

Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal
 

Försäljningsbestämmelser

All försäljning och leverans sker enligt de allmänna leveransbestämmelserna IML 2009 (antagna 2008 av Swedish Labtech) med tillägg av vid var tid tillämpliga försäljningsbestämmelser för VWR, som ska gälla med företräde framför IML 2009 i händelse av motsättning.