Håll mig inloggad?
Avslutas om 
Din inloggning har gått ut
Din session har gått ut. För din säkerhet har vi loggat ut dig.
Vill du logga in igen?

Anti-OLIG2 Rabbit Polyclonal Antibody

Leverantör: ProSci Inc.
OLIG2 is a basic helix-loop-helix transcription factor which is expressed in oligodendroglial tumors of the brain. OLIG2 is an essential regulator of ventral neuroectodermal progenitor cell fate. It is associated with T-cell acute lymphoblastic leukemia due to a chromosomal translocation t (14;21) (q11.2;q22). OLIG2 might play a role in learning deficits associated with Down syndrome.This gene encodes a basic helix-loop-helix transcription factor which is expressed in oligodendroglial tumors of the brain. The protein is an essential regulator of ventral neuroectodermal progenitor cell fate. The gene is involved in a chromosomal translocation t (14;21) (q11.2;q22) associated with T-cell acute lymphoblastic leukemia. Its chromosomal location is within a region of chromosome 21 which has been suggested to play a role in learning deficits associated with Down syndrome. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

OLIG2 antibody can be used for detection of OLIG2 by ELISA at 1:62500. OLIG2 antibody can be used for detection of OLIG2 by western blot at 1 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: OLIG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat




 

ORDER

Expandera alla / Komprimera alla
Storlek Förp. VWR art.nr Antal
50 µg 50 µG PRSI27-353
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFIKATIONER

Antigen symbol OLIG2
Antigen name Oligodendrocyte Transcription Factor 2
Reaktivitet Rat, Mouse, Human
Conjugation Unconjugated
Clonality Polyclonal
Värd Rabbit
Western blot Yes
ELISA Yes
Antigen synonymer RACK17, basic domain, BHLHB1, helix-loop-helix protein, PRKCBP2, class B, bHLHe19, human protein kinase C-binding protein RACK17, oligodendrocyte-specific bHLH transcription factor 2, 1, class B basic helix-loop-helix protein 1, class E basic helix-loop-helix protein 19, OLIGO2, protein kinase C-binding protein 2, oligodendrocyte transcription factor 2
Immunogen A synthetic peptide corresponding a region of human OLIG2.
Form Lyophilized
Enter Details

Select the article for which you are looking for Certificate

Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping