Anti-CDX2 Rabbit Polyclonal Antibody

Leverantör: ProSci Inc.
PRSI27-354EA 6850 SEK
PRSI27-354
Anti-CDX2 Rabbit Polyclonal Antibody
Antikroppar
CDX2 encodes a protein that plays an important role in gallbladder carcinogenesis with intestinal differentiation. Cdx2 is a highly sensitive marker for Barrett's esophagus.

CDX2 antibody can be used for detection of CDX2 by ELISA at 1:312500. CDX2 antibody can be used for detection of CDX2 by western blot at 0.5 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CDX2
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR