Håll mig inloggad?
Avslutas om 
Din inloggning har gått ut
Din session har gått ut. För din säkerhet har vi loggat ut dig.
Vill du logga in igen?

Anti-CDX4 Rabbit Polyclonal Antibody

Leverantör: ProSci Inc.
CDX are homeodomain transcription factors related to the Drosophila caudal gene. The vertebrate CDX have been implicated in the development of the posterior embryo. Several signaling molecules, notably retinoic acid (RA) and members of the Wnt (wingless) and fibroblast growth factor (FGF) families, are also implicated in patterning of the posterior vertebrate embryo. CDX family is the target of Wnt, RA and FGF signaling, suggesting that CDX factors act to convey the activity of these signaling molecules to Hox genes.

CDX4 antibody can be used for detection of CDX4 by ELISA at 1:312500. CDX4 antibody can be used for detection of CDX4 by western blot at 1.0 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CDX4
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:




 

ORDER

Expandera alla / Komprimera alla
Storlek Förp. VWR art.nr Antal
50 µg 50 µG PRSI27-356
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFIKATIONER

Antigen symbol CDX4
Antigen name Caudal type Homeobox 4
Reaktivitet Dog, Human
Conjugation Unconjugated
Clonality Polyclonal
Värd Rabbit
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Antigen synonymer Homeobox protein CDX-4., CDX4_HUMAN, Caudal-type homeobox protein 4, CDX4
Immunogen A synthetic peptide corresponding a region of human CDX4.
Form Lyophilized
Enter Details

Select the article for which you are looking for Certificate

Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal