Anti-KLF1 Rabbit Polyclonal Antibody

Leverantör: ProSci Inc.
PRSI27-358EA 6850 SEK
PRSI27-358
Anti-KLF1 Rabbit Polyclonal Antibody
Antikroppar
KLF1 is a transcription factor, originally identified in this laboratory, which plays a crucial role as a transcriptional activator at the adult beta-globin locus.

KLF1 antibody can be used for detection of KLF1 by ELISA at 1:1562500. KLF1 antibody can be used for detection of KLF1 by western blot at 0.5 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: KLF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR