Anti-TCFL5 Rabbit Polyclonal Antibody

Leverantör: ProSci Inc.
PRSI27-359EA 6850 SEK
PRSI27-359
Anti-TCFL5 Rabbit Polyclonal Antibody
Antikroppar
TCFL5 is a new bHLH transcription factor that negatively regulates upstream transcription factor-dependent transcription.

TCFL5 antibody can be used for detection of TCFL5 by ELISA at 1:312500. TCFL5 antibody can be used for detection of TCFL5 by western blot at 2.0/mL.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TCFL5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR