Anti-SALF Rabbit Polyclonal Antibody

Leverantör: ProSci Inc.
PRSI27-362EA 6250 SEK
PRSI27-362
Anti-SALF Rabbit Polyclonal Antibody
Antikroppar
The SALF mRNA is an infrequent but naturally occurring co-transcribed product of the neighboring SBLF and ALF genes. This rare transcript encodes a fusion protein composed of greater than 95% each of the individual elements, stoned B-like factor (SBLF) and TFIIA-alpha/beta-like factor (ALF). The significance of this co-transcribed mRNA and the function of its protein product have not yet been determined.

SALF antibody can be used for detection of SALF by ELISA at 1:62500. SALF antibody can be used for detection of SALF by western blot at 5 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SALF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR