Anti-SP7 Rabbit Polyclonal Antibody

Leverantör: ProSci Inc.
PRSI27-377EA 6850 SEK
PRSI27-377
Anti-SP7 Rabbit Polyclonal Antibody
Antikroppar
SP7 is a C2H2-type zinc finger transcription factor of the SP gene family and a putative master regulator of bone cell differentiation.

SP7 antibody can be used for detection of SP7 by ELISA at 1:62500. SP7 antibody can be used for detection of SP7 by western blot at 0.125 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR