Håll mig inloggad?
Avslutas om 
Din inloggning har gått ut
Din session har gått ut. För din säkerhet har vi loggat ut dig.
Vill du logga in igen?

Anti-NOTCH3 Goat Polyclonal Antibody

Leverantör: ProSci Inc.
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 60+70 kDa bands in human brain lysates after 1 μg/mL antibody staining.1. The 60+70 kDa bands were successfully blocked by incubation with the immunizing peptide. Immunohistochemistry: In paraffin embedded human hippocampus shows select nuclear staining and weaker staining in different comparments of the neuronal cell bodies. Recommended concentration, 4-6 μg/mL.

NOTCH3 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

NOTCH3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NOTCH3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human




 

ORDER

Expandera alla / Komprimera alla
Storlek Förp. VWR art.nr Antal
100 µg 100 µG PRSI45-978
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFIKATIONER

Antigen symbol NOTCH3
Antigen name Notch 3
Reaktivitet Human
Conjugation Unconjugated
Clonality Polyclonal
Värd Goat
Western blot Yes
ELISA Yes
Koncentration 500 ug/mL
Immunogen NOTCH3 antibody was raised against a 13 amino acid synthetic peptide near the C-Terminus of NOTCH3.
Form Liquid
Enter Details

Select the article for which you are looking for Certificate

Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal