Håll mig inloggad?
Avslutas om 
Din inloggning har gått ut
Din session har gått ut. För din säkerhet har vi loggat ut dig.
Vill du logga in igen?

Anti-NQO1 Goat Polyclonal Antibody

Leverantör: ProSci Inc.
ELISA: Antibody detection limit dilution 1:64000. Western Blot: Approximately 30 kDa band observed in human kidney and lung lysates (calculated MW of 30.9 kDa according to NP_000894.1). Recommended concentration: 0.03-0.1 μg/mL.

NQO1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

NQO1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NQO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Dog, Human, Mouse




 

ORDER

Expandera alla / Komprimera alla
Storlek Förp. VWR art.nr Antal
100 µg 100 µG PRSI45-981
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFIKATIONER

Antigen symbol NQO1
Antigen name NAD(P)H Dehydrogenase, Quinone 1
Reaktivitet Human, Dog, Mouse
Conjugation Unconjugated
Clonality Polyclonal
Värd Goat
Western blot Yes
ELISA Yes
Antigen synonymer NMOR1, NMORI, diaphorase (NADH/NADPH) (cytochrome b-5 reductase), azoreductase, DHQU, NAD(P)H:menadione oxidoreductase 1, NAD(P)H menadione oxidoreductase 1, dioxin-inducible 1, dioxin-ind, diaphorase (NADH/NADPH), QR1, DTD, NAD(P)H dehydrogenase, NQO1, dioxin-inducible, quinone 1, DIA4, NAD(P)H:Quinone acceptor oxidoreductase type 1, DT-diaphorase, NAD(P)H:quinone oxireductase, diaphorase-4
Koncentration 500 ug/mL
Immunogen NQO1 antibody was raised against a 12 amino acid synthetic peptide near the C-Terminus of NQO1.
Form Liquid
Enter Details

Select the article for which you are looking for Certificate

Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping