Håll mig inloggad?
Avslutas om 
Din inloggning har gått ut
Din session har gått ut. För din säkerhet har vi loggat ut dig.
Vill du logga in igen?

Anti-NRAS Goat Polyclonal Antibody

Leverantör: ProSci Inc.
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 21 kDa band observed in human lysates of cell line A431 (calculated MW of 21.2 kDa according to NP_002515.1). In transfected HEK293 transiently expressing NRAS bands of approximately 21 kDa and 25 kDa are observed. These bands are not observed in the non-transfected HEK293. Recommended concentration: 0.01-0.03 μg/mL. Immunohistochemistry: In paraffin embedded human small intestine shows staining of cytoplasm and membrane of enterocytes. Recommended concentration, 2-3 μg/mL.

NRAS antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

NRAS antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NRAS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Rat




 

ORDER

Expandera alla / Komprimera alla
Storlek Förp. VWR art.nr Antal
100 µg 100 µG PRSI45-982
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFIKATIONER

Antigen symbol NRAS
Antigen name Neuroblastoma RAS viral (v-ras) Oncogene homolog
Reaktivitet Human, Rat
Conjugation Unconjugated
Clonality Polyclonal
Värd Goat
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Antigen synonymer RASN, Transforming protein N-Ras, N-RAS, GTPase Nras, ALPS4, NRAS1, RAS, NS6, Neuroblastoma RAS viral (v-ras) oncogene homolog, Neuroblastoma RAS, NRAS
Koncentration 500 ug/mL
Immunogen NRAS antibody was raised against a 13 amino acid synthetic peptide near the internal region of NRAS.
Form Liquid
Enter Details

Select the article for which you are looking for Certificate

Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping