Håll mig inloggad?
Avslutas om 
Din inloggning har gått ut
Din session har gått ut. För din säkerhet har vi loggat ut dig.
Vill du logga in igen?

Anti-CREB3L1 Goat Polyclonal Antibody

Leverantör: ProSci Inc.
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 40 kDa band in human brain lysates after 0.3 μg/mL antibody staining.0 kDa according to NP_443086.1. The 40 kDa band was successfully blocked by incubation with the immunizing peptide.

OASIS antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

OASIS antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CREB3L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:




 

ORDER

Expandera alla / Komprimera alla
Storlek Förp. VWR art.nr Antal
100 µg 100 µG PRSI45-989
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFIKATIONER

Antigen symbol CREB3L1
Antigen name cAMP responsive element binding protein 3-like 1
Reaktivitet Human
Conjugation Unconjugated
Clonality Polyclonal
Värd Goat
Western blot Yes
ELISA Yes
Antigen synonymer cAMP-responsive element-binding protein 3-like protein 1, Old astrocyte specifically-induced substance, Cyclic AMP-responsive element-binding protein 3-like protein 1, OASIS, Processed cyclic AMP-responsive element-binding protein 3-like protein 1
Koncentration 500 ug/mL
Immunogen OASIS antibody was raised against a 13 amino acid synthetic peptide near the C-Terminus of OASIS.
Form Liquid
Enter Details

Select the article for which you are looking for Certificate

Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal
 

Försäljningsbestämmelser

All försäljning och leverans sker enligt de allmänna leveransbestämmelserna IML 2009 (antagna 2008 av Swedish Labtech) med tillägg av vid var tid tillämpliga försäljningsbestämmelser för VWR, som ska gälla med företräde framför IML 2009 i händelse av motsättning.