Anti-OSBPL11 Goat Polyclonal Antibody
Leverantör: ProSci Inc.
PRSI46-107EA 6425 SEK
PRSI46-107
Anti-OSBPL11 Goat Polyclonal Antibody
Antikroppar
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 85-90 kDa band observed in human brain lysates (predicted MWs of 86 kDa according to NP_073613). A minor band of unknown identity was also consistently observed at 75 kDa . Recommended for use at 1-2 μg/mL.
ORP11 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
ORP11 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: OSBPL11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human