Vill du logga in igen?
Anti-OSBPL7 Goat Polyclonal Antibody
Leverantör: ProSci Inc.
ELISA: Antibody detection limit dilution >1:32,000. Western Blot: Approximately 40 kDa in both Jurkat and mouse spleen extracts at 1 μg/mL.
ORP7 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
ORP7 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: OSBPL7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
ORDER
Expandera alla / Komprimera alla
Storlek Förp. VWR art.nr Antal
100 µg 100 µG PRSI46-111
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFIKATIONER
Antigen symbol OSBPL7
Antigen name Oxysterol Binding Protein-like 7
Reaktivitet Mouse, Human
Conjugation Unconjugated
Clonality Polyclonal
Host Goat
ELISA Yes
Koncentration 500 ug/mL
Immunogen ORP7 antibody was raised against a 13 amino acid synthetic peptide near the C-Terminus of ORP7.
Form Liquid
Enter Details
Select the article for which you are looking for Certificate
Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Benämning Artikel nummer Availability Enhet Ditt pris Price Per Antal