Anti-OSBP1 Goat Polyclonal Antibody
Leverantör: ProSci Inc.
PRSI46-114EA 6425 SEK
PRSI46-114
Anti-OSBP1 Goat Polyclonal Antibody
Antikroppar
ELISA: Antibody detection limit dilution >1:16,000. Western Blot: Approximately 100 kDa band observed in human liver lysates (calculated MW of 89.4 kDa according to NP_002547.1). Recommended concentration: 1-3 μg/mL.
OSBP1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
OSBP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: OSBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human