Vill du logga in igen?
Anti-OSBPL10 Goat Polyclonal Antibody
Leverantör: ProSci Inc.
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 90 kDa band observed in human duodenum and human testis lysates (calculated MW of 84.0 kDa according to NP_060254.2). Recommended concentration: 1-3 μg/mL.
OSBPL10 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
OSBPL10 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: OSBPL10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
ORDER
Expandera alla / Komprimera alla
Storlek Förp. VWR art.nr Antal
100 µg 100 µG PRSI46-116
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFIKATIONER
Antigen symbol OSBPL10
Antigen name Oxysterol Binding Protein-like 10
Reaktivitet Human
Conjugation Unconjugated
Clonality Polyclonal
Värd Goat
Western blot Yes
ELISA Yes
Antigen synonymer oxysterol-binding protein-like protein 10, OTTHUMP00000161257, FLJ20363, oxysterol binding protein-like 10, OSBP-related protein 10, OSBPL10, oxysterol-binding protein-related protein 10, HGNC:16395, ORP10, OSBP9
Koncentration 500 ug/mL
Immunogen OSBPL10 antibody was raised against a 14 amino acid synthetic peptide near the internal region of OSBPL10.
Form Liquid
Enter Details
Select the article for which you are looking for Certificate
Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal