Software upgrade kit for Thermo Scientific™ E1-ClipTip™ pipettes
Leverantör: Thermo Scientific
LABS2216470EA 1425 SEK
LABS2216470
Software upgrade kit for Thermo Scientific™ E1-ClipTip™ pipettes
This software kit enables customers with original E1-ClipTip™ electronic pipettes (software version 1.27 or earlier) to connect to the My Pipette™ Creator App via USB cable.
The My Pipette™ Creator App allows users to program their pipettes from their PC and to download ready-made, customised protocols from the cloud. Transferring programs to the E1-ClipTip pipette can be done via micro USB cord included in the E1-ClipTip Software Upgrade Kit.
Leveransinformation: The kit includes: USB cable, license code and instructions for the software upgrade and how to connect to the My Pipette Creator App.