VWR® SFR™ Agarose
Leverantör: Avantor
VWR®
J234-250GEA 11900 SEK
J234-250G J234-100G J234-25G
VWR® SFR™ Agarose
Elektroforesgeler Agaros och akylamid
Agarose SFR™ (Super Fine Resolution) is a high resolution sieving agarose with excellent clarity. DNA bands differing in size by 2% can be resolved in the range of 200 to 1000 bp.
  • Super fine resolution - resolve 238 and 242 bp bands
  • Low melting point
  • Nuclease and protease-free
This agarose is suitable for the analysis of AFLPs (Amplified Fragment Length Polymorphisms), STRs (Short Tandem Repeats) and tetranucleotide repeats. The low melting temperature of Agarose SFR™ makes it an excellent medium for analytical and preparative electrophoresis.