3-Chlorosalicylic acid

Leverantör: Molekula

MOLEM22382330EA 0 SEK
MOLEM22382330
3-Chlorosalicylic acid
3-Chlorosalicylic acid
Formel: C₇H₅ClO₃
MW: 172,57 g/mol
Kokpunkt: 320 °C (1013 hPa)
Smältpunkt: 183…183,5 °C
MDL Number: MFCD00010390
CAS nummer: 1829-32-9
EINECS: 217-379-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR