2-Metylhydrokinon
Leverantör: Molekula
|
Varning
|
MOLEM39846130EA
0
SEK
MOLEM39846130
2-Metylhydrokinon
2-Metylhydrokinon
Formel:
C₇H₈O₂ MW: 124,14 g/mol Kokpunkt: 285 °C (1013 hPa) Smältpunkt: 123…126 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00002345 CAS nummer: 95-71-6 EINECS: 202-443-7 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...