HyClone™ HyQ Soy Hydrolysate
Leverantör: HyClone products (Cytiva)
HyClone™
HYCLSH30357.01EA 1575 SEK
HYCLSH30357.01 HYCLSH30357.02
HyClone™ HyQ Soy Hydrolysate
Cellodlingsmedium
HyClone™ HyQ Soy Hydrolysate solution is a plant-derived protein hydrolysate used with animal protein-free cell culture medium.
Regulatory pressures to remove animal-derived components from products intended for human therapeutic use may pose a challenge to some biopharmaceutical and biological manufacturers. This issue is addressed by providing a high-performance line of serum-free media platforms and offering a portfolio of nutritional and growth supplements.