Direct methanol fuel cell (DMFC) screener membrane electrode assembly
45364.KSEA 5500 SEK
ALFA45364.KS
Direct methanol fuel cell (DMFC) screener membrane electrode assembly
  • MEA-5 layer
  • Plus membrane
  • Active area of 25 cm²
Stored at room temperature
Form: fuel cell component