CLASSIC XL safety cabinet with wing doors for storing flammable liquids provides Type 90 the highest level of fire protection. The integral smart assistance system with touch display monitors exhaust air to prevent an explosive atmosphere forming and minimise health risks, while door sensors check the door status (opened/closed) according to the requirements of DIN EN 14470-1.
- Wing doors remain open in any position allowing convenient use
- Thermocouple ensures doors automatically close in case of a fire
- Cylinder lock for security
- Exhaust air connection socket NW 75 and earthing connection on the cabinet roof (conductively connected with the carcass)
- Interior fittings are conductively connected to the carcass
- Visual inspection of the ventilation gate valve
- Adjustable feet for levelling
- 85 mm high set-back base
Outer carcass made from sheet steel, powder coated in RAL 7035 (light grey), the wing doors that are made from sheet steel, powder coated in RAL 1018 (zinc yellow) and the inner carcass made of high quality decor panels in RAL 7035 (light grey).
Smart assistance system 1 consists of smart control with touch display which includes ventilation monitoring sensor and two door status sensors. Control unit has a cage made of sheet steel in silver with sparkling effect plug-in access to the power supply via IEC-60320 C13 appliance-side connector.
Smart assistance system 2 consists of smart control with touch display including GSM module, ventilation monitoring sensor, two door status sensors and a temperature sensor. Control unit has a cage made of sheet steel in silver with sparkling effect plug-in access to the power supply via IEC-60320 C13 appliance-side connector.
Certifikat: Conforms with DIN EN 14470-1, DIN EN 14727, TRbF 30, DIN EN 60079-10, TRGS 510, EK5/AK4 09-10:2009 and GS certified.
Observera: Heavy and bulky, delivery is not included – please contact your local VWR sales office for a delivery price specific to your location.