Pesticide reference standard, Benoxacor, PESTANAL®, analytical standard, Supelco®
Leverantör: Merck
PESTANAL®
46001-50MGEA 766 SEK
46001-50MG
Pesticide reference standard, Benoxacor, PESTANAL®, analytical standard, Supelco®
Standarder Organic Standards
Benoxacor may be used as a reference standard in the determination of benoxacor in wheat and soil samples using gas chromatography (GC) and high-performance liquid chromatography (HPLC).
Benoxacor is a dichloroacetamide herbicide safener, It is used to protect corn against injury from metolachlor, its role as a safener involves inducing the enzymatic mechanism of chloroacetanilide detoxification in plants.