Du sökte efter: Andningsskydd, fläktassisterade
70  results were found
SearchResultCount:"70"
Sort Results
Visa lista Easy View (ny)
Ge feedback på sökresultat
Artikel nummer : (MMMAS212)
Leverantör: 3M
Beskrivning: Regulator Assembly S-212 is a replacement part for use with Versaflo™ Supplied Air System S-200+.
UOM: 1 * 1 Items
Artikel nummer : (MMMAM927)
Leverantör: 3M
Beskrivning: The Versaflo™ coated visor M-927 is a clear, polycarbonate replacement visor designed for use with 3M™ Versaflo™ M-Series Headtops.
UOM: 1 * 1 Items
Leverantör: JSP LTD
Beskrivning: Dust and half masks commonly fail to do the job they are intended to do because of poor fitting and care by the user. Qualitative Fit Testing can be u...
Leverantör: 3M
Beskrivning: The Versaflo™ Forehead Comfort Pads are designed for use with Versaflo™ S-Series head covers and hoods.
Leverantör: 3M
Beskrivning: Versaflo™ S-series hoods and headcovers feature adjustable suspensions (most models) and soft comfort pads that provide excellent fit, stability and t...
Leverantör: 3M
Beskrivning: These air cooling and heating regulators are ideal to individually cool or heat the user's incoming supplied air by up to 28 °C. A good choice for wor...
Leverantör: 3M
Beskrivning: The 3M™ S-series headcovers offer head and face protection, as well as providing eye and face protection from liquid splash and low energy impact (gra...
Leverantör: 3M
Beskrivning: Aircare™ Filtration Units provide high-quality air from an external source. Removing water and oil droplets, solid particles, and odors, as well as co...
Leverantör: JSP LTD
Beskrivning: The PowerCap® Active™ is a lightweight positive pressure respirator with eye and face protection, as well as EN812 bumpcap protection. Designed to pro...
Leverantör: 3M
Beskrivning: The Versaflo™ Belts are specially designed to help users maintain comfort, and replace worn or damaged parts. These belts are designed for use with th...
Leverantör: JSP LTD
Beskrivning: A fully integrated TH3 powered air respirator offering four of the highest levels of protection in one intuitive head-mounted unit. The system provide...
Artikel nummer : (MMMAS222)
Leverantör: 3M
Beskrivning: Breathing Tube S-222 is a replacement breathing tube for use with 3M™ Versaflo™ Supplied Air System S-200+.
UOM: 1 * 1 Items
Artikel nummer : (MMMATR-971)
Leverantör: 3M
Beskrivning: Versaflo™ Air Flow Indicator TR-971 is designed to confirm the manufacturer's design minimum air flow rate of Versaflo™ Powered Air Turbo is being exc...
UOM: 1 * 1 Items
Artikel nummer : (MMMAM-960)
Leverantör: 3M
Beskrivning: Versaflo™ Visor Pivot Kit M-960 is a replacement part designed for use with 3M™ Versaflo™ M-Series Headtops.
UOM: 1 * 1 Items
Artikel nummer : (MMMATR362E)
Leverantör: 3M
Beskrivning: Versaflo™ Spark Arrestor TR-362 is an accessory designed to protect the filter and turbo in Versaflo™ Powered Air Turbo TR-300+.
UOM: 1 * 1 Items
Leverantör: 3M
Beskrivning: The Versaflo™ powered air Turbo TR-600 is a belt-mounted air purifying device. When combined with one of the 3M approved helmets, it forms a power-ass...
Fråga efter pris
Lager för denna produkt är begränsat, men kan vara tillgänglig i ett lager nära dig. Se till att du är inloggad på hemsidan så att tillgängligt lager kan visas. Om call fortfarande visas och du behöver hjälp, ring oss på 08-621 34 20.
Lager för denna produkt är begränsat, men kan vara tillgänglig i ett lager nära dig. Se till att du är inloggad på hemsidan så att tillgängligt lager kan visas. Om call fortfarande visas och du behöver hjälp, ring oss på 08-621 34 20.
VWR International AB säljer bara etanol till skattebefriade kunder. Denna produkt ä reglerad enligt svensk lagstiftning. Om ytterligare information krävs av dig kommer du att kontaktas via e-post.
VWR International AB säljer bara etanol till skattebefriade kunder.
Denna produkt ä reglerad enligt svensk lagstiftning. Om ytterligare information krävs av dig kommer du att kontaktas via email.
Produkten har blockerats av din organisation. Vänligen kontakta din inköpsavdelning för mer information.
Lösenordet och säkerhetsfrågan associerade med ditt Kundnummer har gått ut. Vänligen ändra ditt lösenord och din säkerhetsfråga.
Denna produkt är inte längre tillgänglig. Alternativ kan vara tillgängliga genom att söka med VWR art.nr ovan. Om du behöver ytterligare hjälp, ring VWR Kundservice på 08-621 34 20.
1 - 16 of 70
no targeter for Bottom