Du sökte efter: Pappershanddukar
21  results were found
SearchResultCount:"21"
Sort Results
Visa lista Easy View (ny)
Ge feedback på sökresultat
Leverantör: KIMBERLY CLARK
Beskrivning: Disposable hand towels made from AIRFLEX*, a voluminous, textile-like fabric, 2 or 3 ply.
Leverantör: KIMBERLY CLARK
Beskrivning: These 1-ply interfolded hand towels are made from strong, highly absorbent AIRFLEX* fabric and supplied in cost-saving ‘compressed’ packs (with 18% mo...
Leverantör: KIMBERLY CLARK
Beskrivning: Disposable hand towels made from AIRFLEX* (a voluminous, textile-like fabric).
Leverantör: KIMBERLY CLARK
Beskrivning: These hand towels are made from strong, highly absorbent AIRFLEX* fabric. In combination with the SLIMROLL* dispenser they are ideal for use in high f...
Artikel nummer : (KIMB6617)
Leverantör: KIMBERLY CLARK
Beskrivning: Designed to help you support washroom hygiene standards in high frequency environments without compromising on quality and value.
UOM: 1 * 5.100 Items
Leverantör: KIMBERLY CLARK
Beskrivning: These 2-ply folded hand towels are made from strong, highly absorbent AIRFLEX* fabric (100% recycled fibres).
Artikel nummer : (115-2168)
Leverantör: KIMBERLY CLARK
Beskrivning: These 2-ply folded hand towels are made from strong, highly absorbent AIRFLEX* fabric.
UOM: 1 * 190 Items
Artikel nummer : (KIMB6781)
Leverantör: KIMBERLY CLARK
Beskrivning: Kleenex® Ultra™ Slimroll™ is the most gentle rolled hand towel perfect for elevated care in premium washrooms. Slimroll™ hand towel pairs with our com...
UOM: 1 * 6 Rulle
Artikel nummer : (KIMB6692)
Leverantör: KIMBERLY CLARK
Beskrivning: These blue hand towel make practical choices without compromising on quality, value and uncompromised clean, every single time.
UOM: 1 * 6 Rulle
Leverantör: KIMBERLY CLARK
Beskrivning: These 1-ply interfolded hand towels are made from strong, highly absorbent AIRFLEX* fabric, safe for food use and supplied in cost-saving ‘compressed’...
Artikel nummer : (KIMB6687)
Leverantör: KIMBERLY CLARK
Beskrivning: These blue hand towel make practical choices without compromising on quality, value and uncompromised clean, every single time.
UOM: 1 * 6 Rulle
Artikel nummer : (KIMB5856)
Leverantör: KIMBERLY CLARK
Beskrivning: Pappershanddukar för engångsbruk av AIRFLEX* (ett tjockt, tygliknande material).
UOM: 1 * 1.760 Items
Artikel nummer : (KIMB6063)
Leverantör: KIMBERLY CLARK
Beskrivning: Embossed paper towel rolls for use with CONTROLOMATIC* dispenser 6963.
UOM: 1 * 6 Rulle
Leverantör: Zellstoff-Vertriebs-GmbH & Co. KG
Beskrivning: These 2 ply tissues dry hands quickly and reliably. Ideal for use in washrooms with high frequencies: municipal facilities, restaurants, medical envir...
Leverantör: Zellstoff-Vertriebs-GmbH & Co. KG
Beskrivning: Engångspappersduk tillverkad i cellulosa, 1, 2 eller 3 skikt.
Leverantör: Zellstoff-Vertriebs-GmbH & Co. KG
Beskrivning: Bomullsvävnad.
Fråga efter pris
Lager för denna produkt är begränsat, men kan vara tillgänglig i ett lager nära dig. Se till att du är inloggad på hemsidan så att tillgängligt lager kan visas. Om call fortfarande visas och du behöver hjälp, ring oss på 08-621 34 20.
Lager för denna produkt är begränsat, men kan vara tillgänglig i ett lager nära dig. Se till att du är inloggad på hemsidan så att tillgängligt lager kan visas. Om call fortfarande visas och du behöver hjälp, ring oss på 08-621 34 20.
VWR International AB säljer bara etanol till skattebefriade kunder. Denna produkt ä reglerad enligt svensk lagstiftning. Om ytterligare information krävs av dig kommer du att kontaktas via e-post.
VWR International AB säljer bara etanol till skattebefriade kunder.
Denna produkt ä reglerad enligt svensk lagstiftning. Om ytterligare information krävs av dig kommer du att kontaktas via email.
Produkten har blockerats av din organisation. Vänligen kontakta din inköpsavdelning för mer information.
Lösenordet och säkerhetsfrågan associerade med ditt Kundnummer har gått ut. Vänligen ändra ditt lösenord och din säkerhetsfråga.
Denna produkt är inte längre tillgänglig. Alternativ kan vara tillgängliga genom att söka med VWR art.nr ovan. Om du behöver ytterligare hjälp, ring VWR Kundservice på 08-621 34 20.
1 - 16 of 21
no targeter for Bottom