Ethyl abietate ≥80.0%

Leverantör: TCI EUROPE

TCIAA0002-25GEA 1425 SEK
TCIAA0002-25G
Ethyl abietate ≥80.0%
Ethyl abietate
Formel: C₂₂H₃₄O₂
MW: 330,51 g/mol
Flampunkt: 178°C
MDL Number: MFCD00028860
CAS nummer: 631-71-0
EINECS: 211-166-0

Order Now

Specifikation testresultat

Appearance Colorless to Yellow to Green clear liquid
Purity (GC) min. 80.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR