3-Bromo-p-anisidine ≥96.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Fara
|
TCIAB3320-1GEA
1275
SEK
TCIAB3320-1G
3-Bromo-p-anisidine ≥96.0% (by GC, titration analysis)
3-Bromo-p-anisidine
Formel:
C₇H₈BrNO MW: 202,0485 g/mol |
MDL Number:
MFCD04094339 CAS nummer: 19056-41-8 UN: 2811 ADR: 6.1,III |
Specifikation testresultat
Appearance | White to Brown powder to crystal |
Purity (GC) | min. 96.0 % |
Purity (Nonaqueous Titration) | min. 96.0 % |
Melting point | 60.0 to 64.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...