4-Phenylbenzhydrazide ≥97.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

TCIAB4350-1GEA 375 SEK
TCIAB4350-1G
4-Phenylbenzhydrazide ≥97.0% (by GC, titration analysis)
4-Phenylbenzhydrazide
Formel: C₁₃H₁₂N₂O
MW: 212,2472 g/mol
MDL Number: MFCD00017078
CAS nummer: 18622-23-6
EINECS: 242-451-8

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 193.0 to 197.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR