Chlorobis(4-methylphenyl)phosphine ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
TCIAC3201-1GEA
1075
SEK
TCIAC3201-1G
Chlorobis(4-methylphenyl)phosphine ≥98.0%
Chlorobis(4-methylphenyl)phosphine
Formel:
C₁₄H₁₄ClP MW: 248,69 g/mol Kokpunkt: 350°C |
MDL Number:
MFCD01630844 CAS nummer: 1019-71-2 UN: 3265 ADR: 8,II |
Specifikation testresultat
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...