(R)-α-(2-Oxiranyl)anisole ≥95.0%
Leverantör: TCI EUROPE
|
Fara
|
TCIAG0409-1GEA
2150
SEK
TCIAG0409-1G
(R)-α-(2-Oxiranyl)anisole ≥95.0%
(R)-α-(2-Oxiranyl)anisole
Formel:
C₉H₁₀O₂ MW: 150,1745 g/mol |
MDL Number:
MFCD06659025 CAS nummer: 71031-02-2 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Specific rotation [a]20/D | -12.0 to -16.0 deg (C=1, EtOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...