Thioacetazone ≥98.0%

Leverantör: TCI EUROPE
Varning

TCIAT3632-250MGEA 725 SEK
TCIAT3632-250MG
Thioacetazone ≥98.0%
Thioacetazone
Formel: C₁₀H₁₂N₄OS
MW: 236,2935 g/mol
MDL Number: MFCD00022157
CAS nummer: 104-06-3
EINECS: 203-170-6

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 22.50 to 24.50 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR