GenSaver™ 2.0 are designed for collection, transport and long-term preservation of DNA from dried matrix spots.
- Long-term preservation of DNA
- High DNA recovery
- Direct amplification and direct STR
- High quality STR and NGS data
- Free of amplifiable human DNA
- DNAses and RNAses free
- Antimicrobial activity
These cards are designed for the collection, transport and storage at ambient temperature of DNA from biological fluids. The fiber-based material of these cards is made of pure absorbent fibers, impregnated with a stabilising chemistry intended to prevent degradation of DNA for long-term ambient preservation.
DNA preserved on GenSaver™ 2.0 cards can be used even after 20 years of ambient storage to generate high quality STR and next generation sequencing data.
GenSaver™ 2.0 is suitable for collection of blood samples and GenSaver™ colour 2.0 is suitable for the collection of colourless biosamples, such as saliva, buccal cells and urine.
These cards also offer additional features, preventing the growth of microorganisms during the long-term ambient storage of DNA.
They are customisable and can be tailored to fully accommodate your biological fluids sampling needs.
Certifikat: Meets ISO 18385:2016