2-(2,6-Dichlorophenyl)thioacetamide
|
Varning
|
APOSOR29606-5GEA
0
SEK
APOSOR29606-5G
APOSOR29606-1G
2-(2,6-Dichlorophenyl)thioacetamide
2-(2,6-Dichlorophenyl)thioacetamide
Formel:
C₈H₇Cl₂NS MW: 220,12 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD00052483 CAS nummer: 17518-49-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...