Du sökte efter: Fotodokumentationssystem
22  results were found
SearchResultCount:"22"
Sort Results
Visa lista Easy View (ny)
Ge feedback på sökresultat
Artikel nummer : (730-1470)
Leverantör: VWR Collection
Beskrivning: The VWR® Basic gel documentation system offers reproducible, high resolution digital images for gel documentation. This entry level system is easy to ...
UOM: 1 * 1 Items
Leverantör: VWR Collection
Beskrivning: VWR® Imager gel documentation and analysis systems are available in configurations suitable for fluorescence and chemiluminescence applications. Both ...
Artikel nummer : (730-1471)
Leverantör: VWR Collection
Beskrivning: The VWR® CHEMI only system is dedicated to chemiluminescence imaging. The system is built for high performance automation and features a next generati...
UOM: 1 * 1 Items
Artikel nummer : (730-0496)
Leverantör: VWR Collection
Beskrivning: SMART5 is a new generation of low cost, integrated imaging system for DNA and protein analysis, also gel documentation.
UOM: 1 * 1 Items
Artikel nummer : (733-2351)
Leverantör: Syngene (a division of the Synoptics group)
Beskrivning: The Syngene G:BOX Chemi XX6 gel documentation system offers optimum performance. Highly sensitive, it is designed for advanced chemiluminescent Wester...
UOM: 1 * 1 Items
Leverantör: Syngene (a division of the Synoptics group)
Beskrivning: The G:BOX mini is a compact, multi application imaging system for accurately imaging mini and midi fluorescence and visible gels, multiplex fluorescen...
Leverantör: Corning
Beskrivning: Axygen® gel documentation systems easily capture publication quality, 16 bit TIFF images. The systems are quick to set up and have an intuitive user i...
Artikel nummer : (SYNOGBOX-F3-LFB)
Leverantör: Syngene (a division of the Synoptics group)
Beskrivning: The Syngene G:BOX-F³ gel imaging system is designed for general fluorescence applications, including DNA and protein analysis. The range of applicatio...
UOM: 1 * 1 Items
Artikel nummer : (SYNOGGNOME-XRQ-NPC)
Leverantör: Syngene (a division of the Synoptics group)
Beskrivning: Photodocumentation System, GeneGnome XRQ, 4 MP, 16 bit system with f0.95 lens, without processor, without monitor
UOM: 1 * 1 Items
Artikel nummer : (MLDVIMAGEXPRESSCON)
Leverantör: Molecular Devices
Beskrivning: Capture large 3D organoid and spheroid images with up to double the speed. The ImageXpress® confocal HT.ai system increases image intensities and thro...
UOM: 1 * 1 Items
Artikel nummer : (SYNOGBOX-CHEMI-XX9)
Leverantör: Syngene (a division of the Synoptics group)
Beskrivning: The Syngene G:BOX Chemi XX9 gel documentation system offers optimum performance. Highly sensitive, it is designed for advanced chemiluminescent Wester...
UOM: 1 * 1 Items
Artikel nummer : (733-2350)
Leverantör: Syngene (a division of the Synoptics group)
Beskrivning: The Syngene G:BOX Chemi XRQ gel imaging system is designed for extended fluorescence applications and chemiluminescent Western blots. The G:BOX Chemi ...
UOM: 1 * 1 Items
Artikel nummer : (PIER62242)
Leverantör: Thermo Fisher Scientific
Beskrivning: This black plastic sample screen is used to cover the transilluminator on the myECL™ imager.
UOM: 1 * 1 Items
Artikel nummer : (PIER62267)
Leverantör: Thermo Fisher Scientific
Beskrivning: Plastic imaging target with printed design; 7×6,5 inches (178×165 mm). This product is a replacement for the myECL™ Imager accessory used for focusing...
UOM: 1 * 1 Items
Leverantör: Thermo Fisher Scientific
Beskrivning: These lamp bulbs with ultraviolet emission are used to replace the one of six pre-installed 306 nm UV bulbs in the myECL™ imager. All six lamps must b...
Artikel nummer : (PIER62240)
Leverantör: Thermo Fisher Scientific
Beskrivning: Thermo Scientific™ 600 to 640 nm bandpass filter is a threaded, 50 mm diameter glass filter. This product is used for the addition of a red filter to ...
UOM: 1 * 1 Items
Fråga efter pris
Lager för denna produkt är begränsat, men kan vara tillgänglig i ett lager nära dig. Se till att du är inloggad på hemsidan så att tillgängligt lager kan visas. Om call fortfarande visas och du behöver hjälp, ring oss på 08-621 34 20.
Lager för denna produkt är begränsat, men kan vara tillgänglig i ett lager nära dig. Se till att du är inloggad på hemsidan så att tillgängligt lager kan visas. Om call fortfarande visas och du behöver hjälp, ring oss på 08-621 34 20.
VWR International AB säljer bara etanol till skattebefriade kunder. Denna produkt ä reglerad enligt svensk lagstiftning. Om ytterligare information krävs av dig kommer du att kontaktas via e-post.
VWR International AB säljer bara etanol till skattebefriade kunder.
Denna produkt ä reglerad enligt svensk lagstiftning. Om ytterligare information krävs av dig kommer du att kontaktas via email.
Produkten har blockerats av din organisation. Vänligen kontakta din inköpsavdelning för mer information.
Lösenordet och säkerhetsfrågan associerade med ditt Kundnummer har gått ut. Vänligen ändra ditt lösenord och din säkerhetsfråga.
Denna produkt är inte längre tillgänglig. Alternativ kan vara tillgängliga genom att söka med VWR art.nr ovan. Om du behöver ytterligare hjälp, ring VWR Kundservice på 08-621 34 20.
1 - 16 of 22
no targeter for Bottom