Du sökte efter: Desinficeringsmedel
28  results were found
SearchResultCount:"28"
Sort Results
Visa lista Easy View (ny)
Ge feedback på sökresultat
Leverantör: CONTEC
Beskrivning: Contec® Sterile IPA is a blend of 70% v/v Isopropanol with 30% water for injection or purified water, this ready to use formulation eliminates mixing,...
Leverantör: CONTEC
Beskrivning: Consisting of 70% v/v Isopropanol blended with highly purified water, this ready to use formulation eliminates mixing and filtration. Used for disinfe...
Leverantör: CONTEC
Beskrivning: Contec HydroPure is a blend of 6% stabilised hydrogen peroxide and water for injection (WFI) or purified water. Contec HydroPure is BPR authorised at ...
Leverantör: CONTEC
Beskrivning: Contec® Sterile CyChlor a blend of stabilised hypochlorous acid in purified water and a broad spectrum disinfectant designed for every day use.
Leverantör: CONTEC
Beskrivning: Blend of hydrogen peroxide and peracetic acid.
Leverantör: CONTEC
Beskrivning: Contec® ProChlor is a ready to use sporicidal biocide based on stabilised hypochlorous acid with purified water and is specifically designed for clean...
Artikel nummer : (BIND1002-0022)
Leverantör: Binder
Beskrivning: deconex® SOLARSEPT is used for disinfection when a short exposure and drying time is required.
UOM: 1 * 500 mL
Leverantör: CONTEC
Beskrivning: Stabilised Hypochlorous acid with purified water. Contec CyChlor is a new broad spectrum disinfectant designed for every day use.
Leverantör: CONTEC
Beskrivning: Contec® HydroPure a blend of 6% stabilised hydrogen peroxide and purified water and leaves little to no residue and is ideal for use on most surfaces....
Artikel nummer : (GOJO32676-02-EEU)
Leverantör: GOJO Industries - Europe Ltd
Beskrivning: Fast acting and fast drying spray with broad spectrum bactericidal, fungicidal, mycobactericidal, and virucidal activity for use on non-porous surface...
UOM: 1 * 2 Items
Leverantör: CONTEC
Beskrivning: Contec ProChlor is a fast acting innovative sporicidal biocide designed for use in pharmaceutical cleanrooms. A blend of hypochlorous acid in purified...
Artikel nummer : (135-1556)
Leverantör: CleanAir by Baker
Beskrivning: Disinfection Kits for Microbiology Safety Cabinets, Class II
UOM: 1 * 1 Items
Leverantör: Diversey
Beskrivning: Ready to use, heavy-duty cleaner for effective removal of lime scale and other deposits from toilet bowls and urinals.
Safer Human and Environmental Health
Sustainable Materials
Sustainable Packaging
Waste Reducing
Artikel nummer : (DIVY100862174)
Leverantör: Diversey
Beskrivning: ClearKlens DE is a ready to use sterile disinfectant with 70% ethanol and 30% WFI (water for injection). It has the broad spectrum of activity against...
UOM: 1 * 6 Flaska
Artikel nummer : (DIVY100941436)
Leverantör: Diversey
Beskrivning: Disinfectant cleaner with AHP technology. Powerful surfactant package delivers excellent cleaning performance including descaling and lime limitation
UOM: 1 * 2 Items
Safer Human and Environmental Health
Artikel nummer : (DIVY7511257)
Leverantör: Diversey
Beskrivning: Divodes FG a detergent disinfectant based on a mixture of propanols is specially formulated for use on surfaces that come into contact with food. It i...
UOM: 1 * 2 Kan
Fråga efter pris
Lager för denna produkt är begränsat, men kan vara tillgänglig i ett lager nära dig. Se till att du är inloggad på hemsidan så att tillgängligt lager kan visas. Om call fortfarande visas och du behöver hjälp, ring oss på 08-621 34 20.
Lager för denna produkt är begränsat, men kan vara tillgänglig i ett lager nära dig. Se till att du är inloggad på hemsidan så att tillgängligt lager kan visas. Om call fortfarande visas och du behöver hjälp, ring oss på 08-621 34 20.
VWR International AB säljer bara etanol till skattebefriade kunder. Denna produkt ä reglerad enligt svensk lagstiftning. Om ytterligare information krävs av dig kommer du att kontaktas via e-post.
VWR International AB säljer bara etanol till skattebefriade kunder.
Denna produkt ä reglerad enligt svensk lagstiftning. Om ytterligare information krävs av dig kommer du att kontaktas via email.
Produkten har blockerats av din organisation. Vänligen kontakta din inköpsavdelning för mer information.
Lösenordet och säkerhetsfrågan associerade med ditt Kundnummer har gått ut. Vänligen ändra ditt lösenord och din säkerhetsfråga.
Denna produkt är inte längre tillgänglig. Alternativ kan vara tillgängliga genom att söka med VWR art.nr ovan. Om du behöver ytterligare hjälp, ring VWR Kundservice på 08-621 34 20.
1 - 16 of 28
no targeter for Bottom