COVID-19 antigen rapid test cassette (saliva)
Leverantör: SureScreen Diagnostics
COVID19AGVCDSEA 3225 SEK
COVID19AGVCDS
COVID-19 antigen rapid test cassette (saliva)
System för klinisk diagnostik Clinical reagents and kits
The COVID-19 Antigen Rapid Test Cassette is a rapid chromatographic immunoassay for the qualitative detection of COVID-19 antigen in saliva. The identification is based on the monoclonal antibodies specific for the Nucleocapsid(N)protein of SARS-CoV-2. It is intended to aid in the rapid differential diagnosis of COVID-19 infections.
For additional information see the Instruction For Use enclosed in the dedicated page of this product.