Diammonium tetrachloropalladate (≥36% Pd)
Leverantör: Thermo Scientific
Fara
11882.06EA 15125 SEK
11882.06 11882.03
Diammonium tetrachloropalladate (≥36% Pd)
Diammonium tetrachloropalladate
Formel: H₈Cl₄N₂Pd
MW: 284,31 g/mol
Smältpunkt: Dec.
Densitet: 2,17
Storage Temperature: Omgivande
MDL Number: MFCD00010884
CAS nummer: 13820-40-1
EINECS: 237-498-6