n-Butyllithium 1.6 mol/l i hexan för syntes, Sigma-Aldrich®
|
Fara
|
M-Clarity™ quality level = MQ200
8.18874.0500EA
1740
SEK
8.18874.0500
n-Butyllithium 1.6 mol/l i hexan för syntes, Sigma-Aldrich®
n-Butyllithium
Formel:
C₄H₉Li MW: 64,06 g/mol Densitet: 0,69 g/cm³ (20 °C) Flampunkt: –22 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00009414 EINECS: 203-698-7 UN: 3394 ADR: 4.2 (4.3),I |
Specifikation testresultat
Solubility | (20 °C) (rigorous decomposition) |
Melting Point | -95 °C |
Boiling Point | 69 °C (1013 hPa) |
Vapor Pressure | 163 hPa (20 °C) |
Flash Point | -22 °C |
Explosion Limit | 1.2 - 7.8 %(V) |
Density | 0.68 g/cm3 (20 °C) |
pH | (H2O, 20 °C) alkaline |
Ignition Point | 240 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...