7-Azaindole

Leverantör: Molekula

MOLEM11947640EA 0 SEK
MOLEM11947640
7-Azaindole
7-Azaindole
Formel: C₇H₆N₂
MW: 118,14 g/mol
Kokpunkt: 384 °C (1013 hPa)
Smältpunkt: 103…106 °C
Densitet: 1,301 g/cm³ (20 °C)
MDL Number: MFCD00005606
CAS nummer: 271-63-6
EINECS: 205-981-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR