Du sökte efter: Filter
VWR har ett brett sortiment inom filterpapper, membranfilter, sprutfilter, glasfiberfilter, kvalitativa och kvantitativa filterpapper. Filtren finns i olika storlekar och former, t.ex runda eller fyrkantiga, veckade eller släta.
VWR har ett brett sortiment inom filterpapper, membranfilter, sprutfilter, glasfiberfilter, kvalitativa och kvantitativa filterpapper. Filtren finns i olika storlekar och former, t.ex runda eller fyrkantiga, veckade eller släta.
811  results were found
SearchResultCount:"811"
Sort Results
Visa lista Easy View (ny)
Ge feedback på sökresultat
Artikel nummer : (23988.)
Leverantör: Merck
Beskrivning: This economy water vapor trap is designed for moisture removal for compressed air service to pneumatically-controlled devices.
UOM: 1 * 1 Each
Artikel nummer : (66884-U)
Leverantör: Merck
Beskrivning: Designed for extracting large volumes of aqueous samples. Suitable for EPA methods and environmental methods.
UOM: 1 * 20 Items
Artikel nummer : (57171.)
Leverantör: Merck
Beskrivning: Solid phase extraction disks provides faster flow rates and exhibits less clogging than PTFE discs for the extraction of organic contaminants from dri...
UOM: 1 * 24 Items
Leverantör: Merck
Beskrivning: Precolumn filter kit has a standard 10-32 fitting and fits direct to most column types without introducing an additional dead volume.
Leverantör: VWR Collection
Beskrivning: Typical applications are clarification, sterile filtration, sample preparation and sterile venting.
Leverantör: VWR Collection
Beskrivning: PS funnel and collection reservoir. Systems for vacuum filtration of aqueous solutions including cell culture media, buffers or other biological fluid...
Sale
Leverantör: Avantor
Beskrivning: These solvent inlet filters are used at the low pressure inlet side of the pump to help protect check valves, injector and column from particulate con...
Leverantör: VWR Collection
Beskrivning: Bindemedelsfria glasfiberfilter med hög flödeshastighet, hög belastningskapacitet, stor temperaturtålighet och utmärkt retention av fällningar.
Leverantör: Avantor
Beskrivning: Placed immediately before the column, the precolumn filter traps particles introduced with the sample. For samples, which may irreversibly absorb onto...
Leverantör: VWR Collection
Beskrivning: These vacuum filter units are very useful in large volume samples separation and purification.
Leverantör: Avantor
Beskrivning: Dessa sprutfilter är speciellt utformade för kromatografiprovberedningstillämpningar och optimerade för att ge de mest konsekventa resultaten med mind...
Sale
Leverantör: VWR Collection
Beskrivning: These units are ideal for minimising unnecessary transfers by filtering directly into the centrifuge tube.
Leverantör: Avantor
Beskrivning: Avantor® ACE® UHPLC pre-column filters made from stainless steel/PEEK provide protection for all UHPLC columns. They offer ultra-low dispersion for ma...
Leverantör: Avantor
Beskrivning: Frit made of 316 grade stainless steel.
Leverantör: Avantor
Beskrivning: These filters are made from 100% PEEK.
Leverantör: Avantor
Beskrivning: These precolumn filters with PEEK body and stainless steel frit provide protection for analytical and microbore columns. They connect directly to the ...
Fråga efter pris
Lager för denna produkt är begränsat, men kan vara tillgänglig i ett lager nära dig. Se till att du är inloggad på hemsidan så att tillgängligt lager kan visas. Om call fortfarande visas och du behöver hjälp, ring oss på 08-621 34 20.
Lager för denna produkt är begränsat, men kan vara tillgänglig i ett lager nära dig. Se till att du är inloggad på hemsidan så att tillgängligt lager kan visas. Om call fortfarande visas och du behöver hjälp, ring oss på 08-621 34 20.
VWR International AB säljer bara etanol till skattebefriade kunder. Denna produkt ä reglerad enligt svensk lagstiftning. Om ytterligare information krävs av dig kommer du att kontaktas via e-post.
VWR International AB säljer bara etanol till skattebefriade kunder.
Denna produkt ä reglerad enligt svensk lagstiftning. Om ytterligare information krävs av dig kommer du att kontaktas via email.
Produkten har blockerats av din organisation. Vänligen kontakta din inköpsavdelning för mer information.
Lösenordet och säkerhetsfrågan associerade med ditt Kundnummer har gått ut. Vänligen ändra ditt lösenord och din säkerhetsfråga.
Denna produkt är inte längre tillgänglig. Alternativ kan vara tillgängliga genom att söka med VWR art.nr ovan. Om du behöver ytterligare hjälp, ring VWR Kundservice på 08-621 34 20.
1 - 16 of 811
no targeter for Bottom