Filter apparatus according to Witt, vacuum-tight
Leverantör: DWK Life Sciences
511-0135EA 8100 SEK
511-0135 511-4211 511-4222 511-4210 511-4220
Filter apparatus according to Witt, vacuum-tight
Trattar Filtertrattar Fitreringsapparat i glas
DURAN®, borosilikatglas 3.3, klar.
  • Lid with NS socket
  • Lid and apparatus with flat flange
  • Very good chemical and high temperature resistance
  • suitable for use under vaccum
Complete with interchangeable Lid and KECK™ assembly set.