Vill du logga in igen?
Knitted gloves, Tiger Paw® 76-301
Leverantör: Ansell
Tiger Paw®
Made from knitted polyester coated with cotton, Tiger Paw® offers many of the features and benefits of Tri Grip™ at a more economical price. This heavier construction ensures a more robust glove for heavier applications.
  • Seamless construction avoids irritation to fingers caused by seams
  • Elastic wrist ensures glove remains in constant fixed position
  • Knitted construction allows the air to circulate thus reducing perspiration
  • PVC polka dots provide a secure grip without loss of dexterity
  • Machine washable at 40 °C
Certifikat: EN 388:2016 (214XB), Cat. II
ORDER
Expandera alla / Komprimera alla
Glove size Färg Längd Förp. Förp. VWR art.nr Antal
7 Beige/blå 230 - 270 mm 12 pairs/bag 12 PAIR ANSE76-301/7P
8 Beige/blå 230 - 270 mm 12 pairs/bag 12 PAIR 112-0299
9 Beige/blå 230 - 270 mm 12 pairs/bag 12 PAIR 112-0300
10 Beige/blå 230 - 270 mm 12 pairs/bag 12 PAIR 112-3606
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFIKATIONER
Modell 76-301
Ej handspecifika/handspecifika Handspecifika
Engångs/Återanvändbar Återanvändbar
Fodermaterial Polyester/bomull
Yta Dot coating on palm
Gauge 13
Kragmodell Stickad mudd
EN standard EN 388:2016 (214XB)
Enter Details
Select the article for which you are looking for Certificate
Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal