Du sökte efter: Centrifuger
VWR ebjuder ett brett urval centrifuger. Golvcentrifuger, bordcentrifuger, mikrocentrifuger, centrifuger med eller utan kyla. Olika modeller har olika möjligheter, kontkata oss för att få hjälp att hitta den modell och den rotor som passar dina behov.
VWR ebjuder ett brett urval centrifuger. Golvcentrifuger, bordcentrifuger, mikrocentrifuger, centrifuger med eller utan kyla. Olika modeller har olika möjligheter, kontkata oss för att få hjälp att hitta den modell och den rotor som passar dina behov.
240  results were found
SearchResultCount:"240"
Sort Results
Visa lista Easy View (ny)
Ge feedback på sökresultat
Leverantör: VWR Collection
Beskrivning: VWR® MiniStar silverline, Rotor för 4 st 8x0,2 ml strips, För: MiniStar silverline
Leverantör: VWR Collection
Beskrivning: The compact Micro Star 30 ventilated centrifuge is an economic, versatile, easy to use cross-over centrifuge which combines the features of a high spe...
Leverantör: VWR Collection
Beskrivning: These versatile centrifuges are ideal for a range of routine applications in pharmaceutical and biotechnology industries, as well as for academic rese...
Leverantör: VWR Collection
Beskrivning: VWR®, Adapter 5 ml blood tubes, 13×75 mm (piece), for angle rotor RA 8/15, max. RCF 3566 ×g
Leverantör: VWR Collection
Beskrivning: These small bench top centrifuges are ideal for research and clinical applications in pharmaceutical and biotechnical industries, as well as for acade...
Sale
Leverantör: VWR Collection
Beskrivning: These versatile centrifuges are ideal for a range of routine applications in pharmaceutical and biotechnology industries, as well as for academic rese...
Leverantör: VWR Collection
Beskrivning: VWR®, Rotor for 10×1,8 ml cryotubes, max. RCF 12300 ×g, För: MicroStar 12
Leverantör: VWR Collection
Beskrivning: These personal centrifuges have a fixed speed of 6000 min⁻¹/2000×g, and are designed for applications requiring relatively low g-forces, such as micro...
Sale
Artikel nummer : (521-3345)
Leverantör: VWR Collection
Beskrivning: The CompactStar CS8 is a small ventilated bench top centrifuge designed for the separation of various samples in the fields of chemistry, life science...
UOM: 1 * 1 Items
Leverantör: VWR Collection
Beskrivning: The compact Micro Star 30R refrigerated centrifuge is an economic, versatile and easy to use cross-over centrifuge which combines the features of a hi...
Leverantör: VWR Collection
Beskrivning: Micro Star 21 and 21R microcentrifuges combine power, versatility and convenience in a safe, compact, easy to use laboratory instrument. Both the vent...
Artikel nummer : (521-2849)
Leverantör: VWR Collection
Beskrivning: Ideal for drying microscope slides. It can also be used in a cold room.
UOM: 1 * 1 Items
Leverantör: VWR Collection
Beskrivning: Replacement tube rotor, quick release, 8×1,5/2,0 ml tubes, for MiniStar blueline centrifuge
Artikel nummer : (521-1651)
Leverantör: VWR Collection
Beskrivning: En snabb och lättanvänd centrifug med utmärkt prestanda, kompakt.
UOM: 1 * 1 Items
Leverantör: Thermo Scientific
Beskrivning: Carriers for microplates, 4×6 standards/4×2 deep well, 86×128 mm, for TX-1000 rotor, För: Megafuge® 40/40R, Multifuge X3/X3R
Leverantör: SORVALL
Beskrivning: Swinging bucket rotor, 8×2000 ml, 3900 min-1, 5374 ×g, incl. oval buckets and 2000 ml bottles, För: Sorvall™ BIOS 16
Fråga efter pris
Lager för denna produkt är begränsat, men kan vara tillgänglig i ett lager nära dig. Se till att du är inloggad på hemsidan så att tillgängligt lager kan visas. Om call fortfarande visas och du behöver hjälp, ring oss på 08-621 34 20.
Lager för denna produkt är begränsat, men kan vara tillgänglig i ett lager nära dig. Se till att du är inloggad på hemsidan så att tillgängligt lager kan visas. Om call fortfarande visas och du behöver hjälp, ring oss på 08-621 34 20.
VWR International AB säljer bara etanol till skattebefriade kunder. Denna produkt ä reglerad enligt svensk lagstiftning. Om ytterligare information krävs av dig kommer du att kontaktas via e-post.
VWR International AB säljer bara etanol till skattebefriade kunder.
Denna produkt ä reglerad enligt svensk lagstiftning. Om ytterligare information krävs av dig kommer du att kontaktas via email.
Produkten har blockerats av din organisation. Vänligen kontakta din inköpsavdelning för mer information.
Lösenordet och säkerhetsfrågan associerade med ditt Kundnummer har gått ut. Vänligen ändra ditt lösenord och din säkerhetsfråga.
Denna produkt är inte längre tillgänglig. Alternativ kan vara tillgängliga genom att söka med VWR art.nr ovan. Om du behöver ytterligare hjälp, ring VWR Kundservice på 08-621 34 20.
1 - 16 of 240
no targeter for Bottom