Du sökte efter: Konduktivitetsmätare
43  results were found
SearchResultCount:"43"
Sort Results
Visa lista Easy View (ny)
Ge feedback på sökresultat
Artikel nummer : (663-0160)
Leverantör: VWR Collection
Beskrivning: Conductivity meter, VWR®, EC30, Tester, Konduktivitet: 0 - 1990 µS/cm, TDS: 0 - 10 ppt, 0...+100 °C, Åtta LR44-batterier, IP 67
UOM: 1 * 1 Items
Leverantör: VWR Collection
Beskrivning: These portable, economical meters can simultaneously measure conductivity, specific conductance, salinity, TDS (Total Dissolved Solids) and temperatur...
Sale
Leverantör: VWR Collection
Beskrivning: User friendly instrument distinguished by its large 127 mm colour graphic touch screen. The meter can be used for accurate measurements of conductivit...
Artikel nummer : (663-0348)
Leverantör: VWR Collection
Beskrivning: This precise device, used for measuring conductivity up to 100 mS/cm, is supplied with a durable wide range graphite measuring cell for universal use ...
UOM: 1 * 1 Items
Artikel nummer : (663-0234)
Leverantör: VWR Collection
Beskrivning: This user friendly portable instrument with robust IP 67 rated housing provides high resolution and accuracy for precise measurements in field applica...
UOM: 1 * 1 SET
Artikel nummer : (663-0349)
Leverantör: VWR Collection
Beskrivning: This high resolution device for measuring the conductivity up to 100 μS/cm is supplied with a stainless steel measuring cell for measurements in pure ...
UOM: 1 * 1 Items
Leverantör: WTW
Beskrivning: Waterproof handheld meters designed for unlimited outside use.
Leverantör: WTW
Beskrivning: Högkvalitativt instrument med avancerad mätteknik vilket gör mätningarna bekvämare och mer tillförlitliga än någonsin förut.
Artikel nummer : (663-0233)
Leverantör: VWR Collection
Beskrivning: This user friendly instrument, with articulated electrode stand and IP 43-rated housing, provides high resolution and accuracy for precise measur...
UOM: 1 * 1 SET
Artikel nummer : (663-0359)
Leverantör: VWR Collection
Beskrivning: These portable, economical meters can simultaneously measure conductivity, specific conductance, salinity, TDS (Total Dissolved Solids) and temperatur...
UOM: 1 * 1 Items
Leverantör: Mettler - Toledo
Beskrivning: These robust meters provide durability and one-handed operation, making them ideally suited for mobile applications in the laboratory, at-line or outd...
Leverantör: Mettler - Toledo
Beskrivning: These bench top instruments provide high quality conductivity measurements with the simple click of a button. Thanks to their intuitive operation, sle...
Leverantör: Mettler - Toledo
Beskrivning: These portable meters provide high quality conductivity measurements with the simple click of a button. The robust and waterproof design makes the Fiv...
Leverantör: Hach
Beskrivning: These testers with integrated sensor measure conductivity, TDS, salinity and temperature of general water samples. They are waterproof and they float.
Leverantör: Mettler - Toledo
Beskrivning: This convenient, reliable instrument is suitable for complex applications and stringent requirements in regulated markets as well as for routine measu...
Leverantör: Mettler - Toledo
Beskrivning: Robust bench top meter for accurate conductivity determinations in the lab and near production.
Fråga efter pris
Lager för denna produkt är begränsat, men kan vara tillgänglig i ett lager nära dig. Se till att du är inloggad på hemsidan så att tillgängligt lager kan visas. Om call fortfarande visas och du behöver hjälp, ring oss på 08-621 34 20.
Lager för denna produkt är begränsat, men kan vara tillgänglig i ett lager nära dig. Se till att du är inloggad på hemsidan så att tillgängligt lager kan visas. Om call fortfarande visas och du behöver hjälp, ring oss på 08-621 34 20.
VWR International AB säljer bara etanol till skattebefriade kunder. Denna produkt ä reglerad enligt svensk lagstiftning. Om ytterligare information krävs av dig kommer du att kontaktas via e-post.
VWR International AB säljer bara etanol till skattebefriade kunder.
Denna produkt ä reglerad enligt svensk lagstiftning. Om ytterligare information krävs av dig kommer du att kontaktas via email.
Produkten har blockerats av din organisation. Vänligen kontakta din inköpsavdelning för mer information.
Lösenordet och säkerhetsfrågan associerade med ditt Kundnummer har gått ut. Vänligen ändra ditt lösenord och din säkerhetsfråga.
Denna produkt är inte längre tillgänglig. Alternativ kan vara tillgängliga genom att söka med VWR art.nr ovan. Om du behöver ytterligare hjälp, ring VWR Kundservice på 08-621 34 20.
1 - 16 of 43
no targeter for Bottom